Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesp1 Antibody (939826), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB92193G
Description
Mesp1 Monoclonal antibody specifically detects Mesp1 in Human samples. It is validated for Western BlotSpecifications
Mesp1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
939826 | |
Western Blot | |
BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1 | |
E. coli-derived recombinant human MESP-1, Met1-Gln85, Accession # Q9BRJ9 | |
0.1 mL | |
Cardiovascular Biology, Developmental Biology, Stem Cells | |
55897 | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction