Learn More
MESP2 Mouse anti-Human, HRP, Clone: 1B3F9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237424H
Catalog No. NBP237424H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MESP2
1B3F9
Western Blot, ELISA
Purified
MESP2
Purified recombinant fragment of human MESP2 (AA: 37-94 ) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
HRP
PBS with No Preservative
bHLHc6, class C basic helix-loop-helix protein 6, mesoderm posterior 2 homolog (mouse), mesoderm posterior protein 2, SCDO2
Mouse
IgG1
0.1 ml
Primary
145873