Learn More
MESP2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 216481AP
Catalog No. 50-173-3535
Add to cart
Description
Description
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
Specifications
Specifications
MESP2
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q0VG99
bHLHc6, Mesoderm posterior protein 2, MESP2, SCDO2
Rabbit
IgG
150 μL
-20°C
Primary
145873
Western Blot
Unconjugated
MESP2
Liquid
MESP2
MESP2 Fusion Protein Ag16345
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human