Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MESP2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310989100UL
Description
MESP2 Polyclonal specifically detects MESP2 in Mouse samples. It is validated for Western Blot.Specifications
MESP2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
bHLHc6, class C basic helix-loop-helix protein 6, mesoderm posterior 2 homolog (mouse), mesoderm posterior protein 2, SCDO2 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse MESP2 (NP_032615.2). Peptide sequence RRTRPAPAGGQRQSASEREKLRMRTLARALQELRRFLPPSVAPAGQSLTK | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
145873 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction