Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meta-Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
37267-86-0
HO3P
79.979
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid,meta-phosphoric acid,hydrogenphosphonate,m-phosphoric acid,phosphenic acid,unii-mtk99r3uv0,metaphosphoric acid hpo3,x-phosphate-group,phosphonate 1-,hydrogen phosphonate
3084658
OP(=O)=O
37267-86-0
HO3P
79.979
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid,meta-phosphoric acid,hydrogenphosphonate,m-phosphoric acid,phosphenic acid,unii-mtk99r3uv0,metaphosphoric acid hpo3,x-phosphate-group,phosphonate 1-,hydrogen phosphonate
3084658
OP(=O)=O

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Chemical Name or Material meta-Phosphoric Acid