Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Metal Activity Tables with Height Adjustable Legs
Made of metal with 1.25 in. plastic laminate table surface.
$734.00 - $1706.00
Specifications
Adjustable (Height) | 56.5 to 94.6 cm (22.25 to 37.25 in.) |
---|---|
Product Type | Activity Table |
Color | Black |
Material | Metal |
Catalog Number | Mfr. No. | Depth (Metric) | Width (Metric) | Top Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (Metric) | Width (Metric) | Top Style | Price | Quantity & Availability | |||
S42493
|
Diversified Spaces™
X814LBB |
76.2 cm | 152.4 cm | Plastic Laminate |
Each for $829.00
|
|
|||
S42495
|
Diversified Spaces™
X8144 |
76.2 cm | 152.4 cm | Phenolic |
Each for $1,393.00
|
|
|||
S42496
|
Diversified Spaces™
X815LBB |
76.2 cm | 182.8 cm | Plastic Laminate |
Each for $876.00
|
|
|||
S42497
|
Diversified Spaces™
X8152 |
76.2 cm | 182.8 cm | ChemGuard |
Each for $1,006.00
|
|
|||
S42498
|
Diversified Spaces™
X8154 |
76.2 cm | 182.8 cm | Phenolic |
Each for $1,514.00
|
|
|||
S42499
|
Diversified Spaces™
X820LBB |
60.9 cm | 137.1 cm | Plastic Laminate |
Each for $739.00
|
|
|||
S42500
|
Diversified Spaces™
X8202 |
60.9 cm | 137.1 cm | ChemGuard |
Each for $823.00
|
|
|||
S42501
|
Diversified Spaces™
X8204 |
60.9 cm | 137.1 cm | Phenolic |
Each for $1,131.00
|
|
|||
S42502
|
Diversified Spaces™
X824LBB |
91.4 cm | 182.8 cm | Plastic Laminate |
Each for $988.00
|
|
|||
S42504
|
Diversified Spaces™
X8244 |
91.4 cm | 182.8 cm | Phenolic |
Each for $1,706.00
|
|
|||
S42505
|
Diversified Spaces™
X860LBB |
60.9 cm | 152.4 cm | Plastic Laminate |
Each for $734.00
|
|
|||
S42506
|
Diversified Spaces™
X8602 |
60.9 cm | 152.4 cm | ChemGuard |
Each for $819.00
|
|
|||
S42507
|
Diversified Spaces™
X8604 |
60.9 cm | 152.4 cm | Phenolic |
Each for $1,222.00
|
|
|||
S42508
|
Diversified Spaces™
X890LBB |
106.6 cm | 152.4 cm | Plastic Laminate |
Each for $1,023.00
|
|
|||
S42509
|
Diversified Spaces™
X8902 |
106.6 cm | 152.4 cm | ChemGuard |
Each for $1,153.00
|
|
|||
S42510
|
Diversified Spaces™
X8904 |
106.6 cm | 152.4 cm | Phenolic |
Each for $1,660.00
|
|
|||
Description
- Steel table apron and height adjustable legs
- 1 x 2 in. 16 gauge apron attaches to the surface
- Top surface options include phenolic resin, ChemGuard or plastic laminate
- 1.25 in. Plastic Laminate Table Surface
- Frame include 14 gauge outer tube with 16 gauge insert legs that range from 22.25 to 37.25 in. (adjustable height)
- Legs are 1.25 in. square
Certifications
MAS Certified Green
Compliance
SEFA
Warranty and Services
Limited Lifetime
Specifications
56.5 to 94.6 cm (22.25 to 37.25 in.) | |
Activity Table | |
Black | |
Metal |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title