Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Analytics™ Metal Combination Electrodes

With Plug Head and Connection Cable

Manufacturer:  SI Analytics™ 285102002

 View more versions of this product

Catalog No. 15-177-877


Add to cart

Description

Description

  • Platinum Pole Sensor, 1 mm ø
Specifications

Specifications

Plug Head
Glass
3 M KCl
Cable Sold Separately
Type A Glass
-5°C to +100°C
Platinum Diaphragm
12 mm
Silamid™ Reference System
170 mm
PT 61
Metal Combination Electrode
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit