Learn More
Remco™ Metal Detectable Bench Scrappers
For bench top scrapping needs.
Manufacturer:  Remco™ 6960MD4
 View more versions of this product
Catalog No. 17-932-335
Add to cart
Description
Description
  • Semi-ferrous additive in product allows for use in conjunction with a metal detectable system
  • Lightweight material alternative to stainless steel
  • Available in blue, green, red and gray colors
  • Autoclavable to 250°F/121°C
Specifications
Specifications
Polypropylene, Semi-Ferrous Additive
0.1 x 8.3 x 5.1 in.
-30°C to +80°C
Red, Yes
-22°F to +176°F
0.39 lb.