Learn More
Remco™ Metal Detectable Hand Scoops
For material handing in case of high sensitivity.
$31.25 - $43.30
Specifications
Material Polypropylene/ Semi-Ferrous Additive additional material
Sterility Non-sterile
Autoclavable Yes
For Use With (Application) Use in conjunction with a metal detectable system
View More Specs
Products 8
Catalog Number Mfr. No. Color Size Capacity (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Capacity (English) Price Quantity & Availability  
17-932309 View Documents Remco™
6400MD2
Green L x W x H: 11.4 x 4.4 x 3 in. 32 oz. Each for $31.25
Add to cart
17-932311 View Documents Remco™
6400MD4
Red L x W x H: 11.4 x 4.4 x 3 in. 32 oz. Each for $31.25
Add to cart
17-932313 View Documents Remco™
6500MD2
Green L x W x H: 14.8 x 7 x 3.75 in. 82 oz. Each for $43.30
Add to cart
17-932315 View Documents Remco™
6500MD4
Red L x W x H: 14.8 x 7 x 3.75 in. 82 oz. Each for $43.30
Add to cart
17-932316 View Documents Remco™
6500MD5
Gray L x W x H: 14.8 x 7 x 3.75 in. 82 oz. Each for $43.30
Add to cart
17-932-318 View Documents Remco™
6400MD3
Blue L x W x H: 11.4 x 4.4 x 3 in. 32 oz. Each for $31.25
Add to cart
17-932-320 View Documents Remco™
6400MD5
Gray L x W x H: 11.4 x 4.4 x 3 in. 32 oz. Each for $31.25
Add to cart
17-932-322 View Documents Remco™
6500MD3
Blue L x W x H: 14.8 x 7 x 3.75 in. 82 oz. Each for $43.30
Add to cart
Description
Description
  • Semi-ferrous additive in product allows for use in conjunction with a metal detectable system
  • Lightweight material alternative to stainless steel
  • Available in blue, green, red and gray colors
  • Autoclavable to 250°F/121°C
  • Working temperature: -22°F to 176°F(-30°C to 80°C)
Specifications
Specifications
Polypropylene/ Semi-Ferrous Additive additional material
Non-sterile
Yes
Use in conjunction with a metal detectable system