Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Metal-Housed Magnetic Stirring Heating Mantle

Fisherbrand™ Metal-Housed Magnetic Stirring Heating Mantle has a deep hull to minimize damage from spills and allow for maximum heat transfer with minimal risk of flask breakage.

 View more versions of this product

Catalog No. 11-476-341


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Designed to fit boiling & distilling style round bottom flasks
  • Operating temperatures up to 450°C (842°F)
  • Durable plastic-coated chemical resistant metal housing
  • Heating element provides uniform, even heat
  • Built-in power controller and magnetic stirrer
  • Includes stirring pellet
  • Adjustable min/max heating for low/high volume contents
  • Thermally insulated and grounded for safety
  • LED heater-on indicator light
  • 120VAC nominal voltage: 5 ft (1.5 m) long power cord with standard 3-prong plug

Warranty and Services

1-year

Specifications

Specifications

100 mL
Magnetic stirring mantle
120 V
Stirring Heating Mantle
Heating Mantle
Simultaneously heating and stirring contents of a round bottom flask
Built-in control
Round Bottom Flasks
5 ft (1.5 m) long pwer cord with standard 3-prong plug
110 W
450°C max.
Up to 1600 U/min
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit