Learn More
Metal Leading Edge Etchant, Reagents
Metal Leading Edge Etchant, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CM0016001A
| Quantity | 1 L |
|---|
Chemical Identifiers
| 7732-18-5 , 10025-77-1 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Dark Orange/Brown | |
| H2O | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <1 | |
| Non-sterile | |
| Metal Leading Edge Etchant |
| 7732-18-5 , 10025-77-1 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.2 | |
| 1 L | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.