Learn More
Brady™ MetaLabel™ B-486 Metallized Polyester Labels with an Aggressive Adhesive
For use in commercial/industrial applications. Brady B-486 metalized polyester labels with an aggressive adhesive are for use with BMP71 printer.
$52.50 - $100.10
Specifications
Product Type Asset Tracking Label
Material Metallized Polyester (B-486)
Adhesive Type Permanent, Rubber
Description Chemical, Corrosion, Humidity, UV and Weather-Resistant
Certifications/Compliance UL Recognized, CSA Approved
View More Specs
Products 5
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Size (English) Price Quantity & Availability  
19-109-071 View Documents Brady™
M7117486
W x H: 25.4 x 12.7 mm W x H: 1 x 0.5 in. Each for $89.10
Add to cart
19-109-080 View Documents Brady™
M7120486
W x H: 50.8 x 25.4 mm W x H: 2 x 1 in. Each for $52.50
Add to cart
19-109-097 View Documents Brady™
M7129486
W x H: 38.1 x 12.7 mm W x H: 1.5 x 0.5 in. Each for $100.10
Add to cart
19-109-125 View Documents Brady™
M7130486
W x H: 38.1 x 19.1 mm W x H: 1.5 x 0.75 in. Each for $83.00
Add to cart
19-109-139 View Documents Brady™
M7137486
W x H: 76.2 x 48.3 mm W x H: 3 x 1.9 in. Each for $76.00
Add to cart
Specifications
Specifications
Asset Tracking Label
Permanent, Rubber
UL Recognized, CSA Approved
Matte
Asset Tracking, Rating Plates, General Purpose ID
Metallized Polyester (B-486)
Chemical, Corrosion, Humidity, UV and Weather-Resistant
0.0053 in.
BMP71 printer
Light Gray