Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metallothionein-1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30349620UL
Description
Metallothionein-1B Polyclonal specifically detects Metallothionein-1B in Human samples. It is validated for Western Blot.Specifications
Metallothionein-1B | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
4490 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
metallothionein-1B, metallothionein 1B, MT1, MT1Q, MTP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-61 of human Metallothionein-1B (NP_005938.1). | |
20 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction