Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Metalware Set - Superior

Manufacturer:  Eisco™ MTST6/DEL

Catalog No. 04-999-164

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Base retort stand, 25 x 16 cm, 2000 g (CH0654B)
  • Stainless steel rod of 600 mm (CH0659B)
  • Bosshead (CH0663D)
  • Three prong adjustable clamp (CH0676A)
  • Cast iron ring of 50 mm dia. (CH0674A)
  • Cast iron ring of 100 mm dia. (CH0674C)
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit