Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ METAP1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer: Abnova™ H00023173T01
Specifications
Western Blot | |
METAP1 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 30.4kDa | |
Transient overexpression cell lysate was tested with anti-METAP1 antibody (Mfr. No. H00023173-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
293T | |
NM_015143 | |
pCMV-METAP1 full-length | |
100 μL | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title