Learn More
METAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 170401AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-1343
Add to Cart
Description
Description
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintece of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
Specifications
Specifications
METAP2
Polyclonal
Unconjugated
METAP2
MAP 2, MAP2, MetAP 2, METAP2, Methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67eIF2, Peptidase M 2
Rabbit
Antigen Affinity Chromatography
RUO
10988, 56307, 64370
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08663, P38062, P50579
Metap2
METAP2 Fusion Protein Ag10564
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions