Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metaphosphoric acid, Acros Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
37267-86-0,50813-16-6
HO3P
79.979
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid,meta-phosphoric acid,hydrogenphosphonate,m-phosphoric acid,phosphenic acid,unii-mtk99r3uv0,metaphosphoric acid hpo3,x-phosphate-group,phosphonate 1-,hydrogen phosphonate
3084658
OP(=O)=O
37267-86-0,50813-16-6
HO3P
79.979
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid,meta-phosphoric acid,hydrogenphosphonate,m-phosphoric acid,phosphenic acid,unii-mtk99r3uv0,metaphosphoric acid hpo3,x-phosphate-group,phosphonate 1-,hydrogen phosphonate
3084658
OP(=O)=O
Quantity 500g
Linear Formula (HPO3)n
Grade ACS Reagent
Packaging Glass Bottle
Solubility Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol
Chemical Name or Material Metaphosphoric acid