Learn More
MetaSystems™ CL 20q12/20qter Plus DNA Probe
Detects deletions which occur in long arm of chromosome 20
Manufacturer:  Metasystems Group IncSupplier Diversity Partner D5994100OG
Catalog No. 22-200-359
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Two labeled probes: orange and green
Description
Description
  • Orange labeled hybridizes to 20q12 and includes proximal part of PTPRT
  • Green labeled probe hybridizes to q-terminal region of long arm of chromosome 20
Specifications
Specifications
Two labeled probes: orange and green
10 Tests
100μL