Learn More
MetaSystems CL CCND1/IGH DNA Probe
SDP
Designed as dual fusion probe to detect t(11;14)
Manufacturer:  MetaSystems D5966100OG
Catalog No. 22-200-349
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Three labeled probes: orange and green
Description
Description
  • Orange labeled probe spans breakpoints at 11q13 (CCND1)
  • Green labeled probe flanks 14q32 IGH breakpoint region
Specifications
Specifications
100μL
Two labeled probes: orange and green
10 Tests