Learn More
MetaSystems XT ALK BA DNA Probe
SDP
Break apart probe
Manufacturer:  MetaSystems D6001100OG
Catalog No. 22-200-353
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Orange labeled probe hybridizing to proximal region within ALK in 2p23
  • Green labeled probe hybridizing distal to ALK
  • Used for tissue FISH applications
Specifications
Specifications
100μL
Two labeled probes: orange and green
Tissue-FISH applications
MetaSystems' pretreatment kit.
10 Tests