Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metaxin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Metaxin 1
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092577
View Documents
Novus Biologicals
NBP303536100UL
100 μg Each for $320.71
Add to cart
 
NB092578
View Documents
Novus Biologicals
NBP30353620UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Metaxin 1 Polyclonal antibody specifically detects Metaxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

Metaxin 1
Unconjugated
Rabbit
Human, Mouse, Rat
metaxin 1, metaxin-1, MTX, MTXNMitochondrial outer membrane import complex protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 186-435 of human Metaxin 1 (NP_942584.2).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
4580
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit