Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meteorin Rat anti-Mouse, Alexa Fluor™ 750, Clone: 21Q6, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221990AF750

Catalog No. NP221990A70


Add to cart

Description

Description

Meteorin Monoclonal antibody specifically detects Meteorin in Mouse samples. It is validated for ELISA.
Specifications

Specifications

Meteorin
21Q6
ELISA
Conjugated, Purified
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
METRN
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Meteorin. Rat monoclonal anti mouse Meteorin Q8C1Q4.
Protein G purified
Neuroscience
Primary
79006.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit