Learn More
Cole-Parmer Amprobe ACD 1000A Digital Clamp-On Multimeter  DFS Item
Manufacturer:  Cole-Parmer 2000333
With a rugged design that is ideal for industrial environments, this digital clamp-on multimeter features auto and manual ranging, relative zero mode and a data hold. It has an auto power off and large easy-to-read display, too. It is available for sale only in the US and Canada.
Catalog No. 50-630-896