Learn More
Cole-Parmer Balmac Inc. 242 Pocket Vibration Meter; 3 to 1000 Hz  DFS Item
Manufacturer:  Cole-Parmer 9886110
Measure displacement, velocity and acceleration of motors, pumps, and other process systems. Large LCD reads parameters in English units. Hold feature freezes display for 60 seconds.
Catalog No. 50-627-391