Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylamide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
2.5kg
79-39-0
C4H7NO
85.106
MFCD00008018
FQPSGWSUVKBHSU-UHFFFAOYSA-N
methacrylamide,2-methylacrylamide,methacrylic amide,methacrylic acid amide,2-propenamide, 2-methyl,2-methylpropenamide,methacryamide,methylacrylic amide,mhoromer bm801,alpha-methyl acrylic amide
6595
CHEBI:51759
2-methylprop-2-enamide
CC(=C)C(=O)N
79-39-0
C4H7NO
85.106
MFCD00008018
FQPSGWSUVKBHSU-UHFFFAOYSA-N
methacrylamide,2-methylacrylamide,methacrylic amide,methacrylic acid amide,2-propenamide, 2-methyl,2-methylpropenamide,methacryamide,methylacrylic amide,mhoromer bm801,alpha-methyl acrylic amide
6595
CHEBI:51759
2-methylprop-2-enamide
CC(=C)C(=O)N
Water 1% max. (K.F.)
Melting Point 108°C to 112°C
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
Sublimation Point 95°C
Assay Percent Range 98%
Linear Formula H2C=C(CH3)CONH2
Beilstein 02, II, 399
Formula Weight 85.11
Physical Form Crystals
Percent Purity ≥97.5%
Grade Extra Pure
Chemical Name or Material Methacrylamide, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-202-3

RTECSNumber : UC6475000

TSCA : TSCA