Learn More
Sigma Organic Chemistry Methacrylic acid | 100G | 79-41-4 | MFCD00002651 | 0.99
Supplier: Sigma Organic Chemistry 155721100G
Methacrylic acid, 100G
About This Item:
Synonym(s): 2-Methacrylic acid; 2-Methylpropenoic acid
Linear Formula: H2C=C(CH3)COOH
Molecular Weight: 86.09
CAS Number: 79-41-4
MDL Number: MFCD00002651
UNSPSC Code: 12162002
Purity: 0.99
Boiling Point: 163 C (lit.)
Melting Point: 12-16 C (lit.)
Density: 1.015 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.431 (lit.)
Storage: room temp
EINECS Number: 201-204-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.