Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol

Manufacturer: Alfa Aesar 1554630

Quantity 250g

 View more versions of this product

Catalog No. AA1554630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-41-4
    Molecular Formula CH2=C(CH3)CO2H
    Formula Weight 86.09
    MDL Number MFCD00002651
    UN Number UN2531
    Beilstein 1719937
    Merck Index 14,5941
    Synonym 2-Methylacrylic acid
    Chemical Name or Material Methacrylic acid, Stabilized with 250ppm 4-methoxyphenol

    Specifications

    Formula Weight 86.09
    Quantity 250g
    TSCA TSCA
    UN Number UN2531
    Beilstein 1719937
    Merck Index 14,5941
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H302-H312-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immed
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHACRYLIC ACID, STABILIZED
    RTECS Number OZ2975000
    Physical Form Liquid
    Density 1.015
    Molecular Formula CH2=C(CH3)CO2H
    MDL Number MFCD00002651
    Assay Percent Range >99%
    CAS 79-41-4
    Synonym 2-Methylacrylic acid
    Chemical Name or Material Methacrylic acid, Stabilized with 250ppm 4-methoxyphenol
    Assay ≥99%

    GHS H Statement
    H314-H302-H312-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immed

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit