Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methallyl phenyl ether, 96%
Click to view available options
Quantity:
5g
25g
Specifications
Refractive Index | 1.515 |
Density | 0.964 |
Flash Point | 77°C (171°F) |
Boiling Point | 204°C |
Quantity | 25g |
Beilstein | 1933868 |
Formula Weight | 148.21 |
Percent Purity | 96% |
Chemical Name or Material | Methallyl phenyl ether |
Safety and Handling
GHS H Statement
H334-H227-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
May cause an allergic skin reaction.
P210-P261-P272-P280-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P370+P378q-P501c
H227-H317-H334
EINECSNumber : 227-393-3
TSCA : No
Recommended Storage : Ambient temperatures