Learn More
Strem, An Ascensus Company Xantpho PalladacycleG398%500mg
SDP
Supplier:  Strem, An Ascensus Company 460957500MG
Encompass_Preferred
CAS# 1445085-97-1. 500mg. Methanesulfonato[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene][2'-amino-1,1'-biphenyl]palladium(II) dichloromethane adduct, min. 98% [Xantphos Palladacycle Gen. 3]. MFCD25372941. Molecular Weight: 948.35. Molecular Formula: C52H45NO4P2PdS. Color/form: pale yellow pwdr. Strem# 46-0957. http://www.strem.com/catalog/v/46-0957/
Catalog No. 50-138-5537