Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic Acid, 70% Solution in Water, Pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
5L
Packaging:
Glass Bottle
Plastic bottle
75-75-2
CH4O3S
96.1
MFCD00007518
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O
75-75-2
CH4O3S
96.1
MFCD00007518
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O
Quantity 5L
Packaging Plastic bottle
Density 1.3500g/mL
Refractive Index 1.4130 to 1.4150
Melting Point -54.0°C
Specific Gravity 1.35
Flash Point >110°C
Color Undesignated
Linear Formula CH3SO3H
Beilstein 04, 4
Fieser 01,666; 02,270; 04,326; 10,256; 11,321; 12,212; 13,176
Merck Index 15, 6026
Formula Weight 96.1
Physical Form Solution
Percent Purity 69.5 to 70.5%
Grade Pure
Chemical Name or Material Methanesulfonic acid, 70% Solution in Water
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger