Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic Acid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500g
5kg
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O

A grade of chemicals for laboratory use

Flash Point 189.00°C
Specific Gravity 1L = 1.48kg
Quantity 5kg
Formula Weight 96.1
Chemical Name or Material Methanesulfonic Acid