Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic Acid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500g
5kg
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O

A grade of chemicals for laboratory use

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.48kg
Formula Weight 96.1
Quantity 500g
Flash Point 189.00°C
Chemical Name or Material Methanesulfonic Acid