Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic anhydride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
7143-01-3
C2H6O5S2
174.18
IZDROVVXIHRYMH-UHFFFAOYSA-N
methanesulfonic anhydride,methanesulfonic acid, anhydride,mesyl anhydride,mesic anhydride,methanesulphonic anhydride,methane sulfonic acid anhydride,methanesulfonyl methanesulfonate,methane sulfonic anhydride,methanesulfonic acid, 1,1'-anhydride,methanesulfonic acid methylsulfonyl ester
81560
methylsulfonyl methanesulfonate
CS(=O)(=O)OS(=O)(=O)C
7143-01-3
C2H6O5S2
174.18
IZDROVVXIHRYMH-UHFFFAOYSA-N
methanesulfonic anhydride,methanesulfonic acid, anhydride,mesyl anhydride,mesic anhydride,methanesulphonic anhydride,methane sulfonic acid anhydride,methanesulfonyl methanesulfonate,methane sulfonic anhydride,methanesulfonic acid, 1,1'-anhydride,methanesulfonic acid methylsulfonyl ester
81560
methylsulfonyl methanesulfonate
CS(=O)(=O)OS(=O)(=O)C
Color Gray to White
Boiling Point 125.0°C (4.0 mmHg)
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 69.0°C to 70.0°C
Assay Percent Range 97%
Solubility Information Solubility in water: reacts
Formula Weight 174.18
Physical Form Powder, Crystals and/or Chunks
Percent Purity 97%
Chemical Name or Material Methanesulfonic anhydride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 230-442-1