Alfa Aesar™ Methanesulfonyl fluoride, 98%

Manufacturer: Alfa Aesar™ H53460-06

 View more versions of this product

Catalog No. AAH5346006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 558-25-8
    Molecular Formula CH3FO2S
    Formula Weight 98.09g/mol
    MDL Number MFCD00007423
    UN Number UN3389
    Chemical Name or Material Methanesulfonyl fluoride

    Specifications

    Boiling Point 123° to 124°C
    Quantity 5g
    CAS 558-25-8
    Molecular Formula CH3FO2S
    Formula Weight 98.09g/mol
    MDL Number MFCD00007423
    UN Number UN3389
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H330-H314-H318
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
    RTECS Number PB2975000
    TSCA TSCA
    Chemical Name or Material Methanesulfonyl fluoride
    Assay 98%

    GHS H Statement
    H300-H330-H314-H318
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit