Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 83% v/v HPLC, Reagents

Catalog No. 01334734
Change view
Click to view available options
:
Quantity: 4 L
Catalog No.
01-334-734 Quantity: 4 L
1 options

Catalog No. 01-334-734

Supplier: Reagents Holdings Llc CM1230004C

Only null left
Add to Cart
Add to Cart

Methanol, 83% v/v HPLC, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Boiling Point 67°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.856
Percent Purity 83%
Grade Reagent
Chemical Name or Material Methanol, 83% v/v HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.