Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Plastic drum
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
Additional Information Vapor Pressure: 128hPa at 20°C
Assay Percent Range 99+%
Linear Formula CH3OH
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.7910g/mL
Packaging Plastic drum
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 0.005% max.
Solubility Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Specific Gravity 0.791
Water 0.2% max. (K.F.)
Flash Point 12°C
Viscosity 0.55 mPa.s (20°C)
Boiling Point 64.7°C
Color Undesignated
Melting Point -98.0°C
Quantity 5L
Chemical Name or Material Methanol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger