Methanol, Absolute, PHOTREX™ Reagent, J.T.Baker™

High purity solvents characterized for use in UV, visible and IR spectrophotometry

Manufacturer: J.T.Baker 906901

Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL

 View all options for this product

Catalog No. 02-003-354

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 67-56-1
    Molecular Formula CH3OH
    Formula Weight 32.04g/mol
    Chemical Name or Material Methanol, Absolute
    Grade PHOTREX™ Reagent

    Specifications

    Packaging Narrow Mouth Amber Glass Bottle
    Quantity 500mL,500mL
    Specific Gravity 1L = 0.80kg
    Flash Point 12.00°C
    CAS 67-56-1
    Molecular Formula CH3OH
    Formula Weight 32.04g/mol
    Chemical Name or Material Methanol, Absolute
    Grade PHOTREX™ Reagent

    Description

      For Spectrophotometry

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit