Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
Click to view available options
Quantity:
500mL
1L
4L
16L
20L
Packaging:
Poly natural
Ropak™
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Physical Form | Liquid |
Grade | ACS Reagent |
Packaging | Poly natural |
Color | Colorless |
Quantity | 500mL |
Chemical Name or Material | Methanol, Absolute |