Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
Click to view available options
Quantity:
25 mL
500 mL
1 L
4 L
16 L
20 L
Packaging:
Amber Glass Bottle
Hard Pack
Metal Can
Natural Poly Bottle
Poly Natural Bottle
Specifications
Color | Colorless |
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
Percent Purity | ≥99.8% |
Grade | ACS |
Quantity | 1 L |
Packaging | Natural Poly Bottle |
Physical Form | Liquid |
Residue after Evaporation | max. 0.001% |
Show More |
Safety and Handling
ShelfLife | 36 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction