Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Pressure Bottom Poly Bottle

Chemical Identifiers

67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CHEBI:17790
methanol
CO

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 32.04
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 12.00°C (53.6°F)
Quantity 4L
Chemical Name or Material Methanol
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements