Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™
Click to view available options
Quantity:
6 x 1 L
Packaging:
Narrow Mouth Flint
Description
- For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use
Specifications
Melting Point | 113°C to 116°C |
Quantity | 6 x 1 L |
Flash Point | 12.00°C |
Packaging | Narrow Mouth Flint |
Specific Gravity | 1L = 0.79kg |
Formula Weight | 32.04 |
Grade | BAKER ANALYZED™ ULTRA LC/MS |
Chemical Name or Material | Methanol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction