Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T.Baker™

Click to view available options
Quantity:
2X1L
6 x 1L
Packaging:
Narrow Mouth Flint

Chemical Identifiers

67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CHEBI:17790

For cutting-edge LC/MS applications

    For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use
CAS 67-56-1
Molecular Formula CH4O
UN Number 1230
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.042
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04
Packaging Narrow Mouth Flint
Specific Gravity 1L = 0.79kg
Flash Point 12.00°C (53.6°F)
Melting Point 113° to 116°C
Quantity 2X1L
Chemical Name or Material Methanol
Grade BAKER ANALYZED™ ULTRA LC/MS
Show More Show Less

DOTInformation : Class 3: Flammable Liquid