Alfa Aesar™ Methanol-d4, 99.8% (Isotopic)

Manufacturer: Alfa Aesar™ 42318KV

Quantity 10 each

 View more versions of this product

Catalog No. AA42318KV

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 811-98-3
    Formula Weight 36.07
    MDL Number MFCD00044637
    UN Number UN1230
    Chemical Name or Material Methanol-d4, Isotopic

    Specifications

    Formula Weight 36.07
    Packaging Prescored Ampules
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 64.7°C
    MDL Number MFCD00044637
    UN Number UN1230
    Health Hazard 1 Danger
    TSCA TSCA
    Density 0.89g/mL
    Melting Point −98°C
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H225-H300-H310-H330-H370
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
    Quantity 10 each
    CAS 811-98-3
    Odor Mild
    Chemical Name or Material Methanol-d4, Isotopic
    Assay 99.8%

    GHS H Statement
    H225-H300-H310-H330-H370
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit