Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 32.04
Grade BAKER ANALYZED™ LC-MS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 12.00°C
Quantity 4L
Chemical Name or Material Methanol, Filtered through a 0.2 micron filter