Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, For HPLC analysis, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
Hydroxymethane; Methyl hydrate
887
CHEBI:17790
methanol
CO

High purity solvents for use in analytical and preparative separations

Formula Weight 32.04
Physical Form Liquid
Density 0.7918g/cm3
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 12°C
Boiling Point 64.7°C
Color Colorless
Melting Point -97.6°C
Quantity 4L
Chemical Name or Material Methanol, For HPLC analysis
Grade HPLC Grade
Show More Show Less