Methanol (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Used in histology applications

Packaging Steel Pail
Quantity 20L

 View more versions of this product

Catalog No. A433S-20

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 67-56-1
    Physical Form Liquid
    Molecular Formula CH4O
    Linear Formula CH3OH
    Formula Weight 32.04
    MDL Number MFCD00004595
    Synonym Methyl Alcohol
    Chemical Name or Material Methanol
    Grade Histological Grade

    Specifications

    Melting Point −98°C
    Boiling Point 64.7°C
    Formula Weight 32.04
    Quantity 20L
    Flash Point 12°C (53.6°F)
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Colorless
    Viscosity 0.55 cP at 20°C
    Density 1.11 (Air = 1.0)
    Vapor Pressure 128 hPa at 20°C
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

    NFPA

    Health:1
    Flammability:3
    Instability:0
    Identification Pass Test
    Cleanup Symbol Solvent
    Molecular Formula CH4O
    CAS 67-56-1
    Health Hazard 2 DANGER!
    Additional Information Vapor Pressure: 128hPa at 20°C
    Packaging Steel Pail
    Linear Formula CH3OH
    MDL Number MFCD00004595
    ChemAlert Storage Symbol Red
    Synonym Methyl Alcohol
    Chemical Name or Material Methanol
    Grade Histological Grade
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

    NFPA

    Health:1
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit