Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Histological), Fisher Chemical

Used in histology applications

Packaging Aluminum Can
Quantity 4L

 View more versions of this product

Catalog No. A433S-4



  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

67-56-1
CH4O
32.04
Methyl Alcohol
Histological Grade
Liquid
CH3OH
MFCD00004595
Methanol

Specifications

67-56-1
Liquid
CH3OH
MFCD00004595
Methyl Alcohol
Pass Test
128 hPa at 20°C
0.55 cP at 20°C
Colorless
4L
Histological Grade
Vapor Pressure: 128hPa at 20°C
CH4O
32.04
1230
1.11 (Air = 1.0)
Aluminum Can
12°C (53.6°F)
64.7°C
−98°C
Methanol

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit