Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

HPLC mobile phase for analytical and preparative separations

Packaging Glass Bottle, Safe-Cote™
Quantity 1L

 View more versions of this product

Catalog No. A452SK-1



  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

67-56-1
CH4O
32.04
MeOH
≥99.9%
Assay: 99.9% min by GC
CH3OH
MFCD00004595
Methanol
HPLC Grade, Mobile phase for HPLC applications

Specifications

0.025 AU max. at 254nm
Vapor Pressure: 128hPa at 20°C
Assay: 99.9% min by GC
CH3OH
MFCD00004595
MeOH
0.001% max.
1.11 (Air = 1.0)
Pass Test
Pass Test
1.3260 to 1.3300 (at 25°C)
Pass Test
Pass Test
0.0003mEq/g max.
128 hPa at 20°C
0.55 cP at 20°C
64.7°C
−98°C
Methanol
HPLC Grade, Mobile phase for HPLC applications
67-56-1
Liquid
CH4O
32.04
1230
Solubility in water: miscible
HPLC mobile phase for analytical and preparative separations. High UV transmittance and low evaporation residue. Suitable for gradient and isocratic conditions. Filtered through a 0.2μm filter to remove particulates. This product also meets ACS specifications.
Pass Test
Pass Test
Glass Bottle, Safe-Cote™
3ppm max.
0.791
Pass Test
0.0002mEq/g max.
0.1% max.
0.2-micron filtered.
Colorless
1L
≥99.9%

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and eyes. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit