Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Laboratory Reagent, ≥99.6%, Honeywell™

Click to view available options
Quantity:
1L
2.5L
10L
20L
4 x 2.5L
4 x 4L
4 x 5L
160kg
Packaging:
Fiberboard Box
Glass Bottle
Metal Drum
Stainless Steel Can
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO

Laboratory Reagent,≥99.6%

  • ≥95% for general laboratory use.
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.6%
Grade Laboratory Reagent
Density 0.791g/mL at 25°C
Explosion Limit 0.36
Ignition Point 725°F
Packaging Glass Bottle
Refractive Index n20/D 1.329(lit.)
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
Vapor Density 1.11 (vs air)
Flash Point 9°C (48.2°F)
Physical State Liquid
Boiling Point 65°C
Color Colorless
Melting Point -98°C
Quantity 1L
Chemical Name or Material Methanol
Show More Show Less

P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235

H225-H301 + H311 + H331-H370

DOTInformation : class 3 - PG 2 - Methanol

EINECSNumber : 200-659-6

RTECSNumber : PC1400000