Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Low water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Formula Weight 32.04
Grade BakerDRY™
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Specific Gravity 1L = 0.80kg
Flash Point 12.00°C
Quantity 1L
Chemical Name or Material Methanol, Low water